missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Galacton-Plus™ Substrate (100X Concentrate)
Galacton-Plus™ Substrate (100X Concentrate)
£233.00 - £938.00
Specifications
Content And Storage | Store the product at 4°C. |
---|---|
Form | Liquid |
Product Line | Galacton-Plus™ |
Substrate | Beta-Galactosidase |
Substrate Type | Beta-Gal Substrate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12694315
|
Molecular Probes™
T2118 |
2mL |
£233.00
Each |
Estimated Shipment: 16-04-2024
Log in to see stock. |
|||||
12604325
|
Molecular Probes™
T2119 |
4mL |
£418.00
Each |
Estimated Shipment: 16-04-2024
Log in to see stock. |
|||||
12614325
|
Molecular Probes™
T2120 |
10mL |
£938.00
Each |
Estimated Shipment: 16-04-2024
Log in to see stock. |
|||||
Description
The Galacton-Plus™ Substrate is a concentrated chemiluminescent substrate for E.coli β-galactosidase assays.For Research Use Only. Not for use in diagnostics procedures.
Specifications
Store the product at 4°C. | |
Galacton-Plus™ | |
Beta-Gal Substrate | |
Room Temperature |
Liquid | |
Beta-Galactosidase | |
Bottle(s) |
For Research Use Only. Not for use in diagnostic procedures.