missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Galacton-Plus™ Substrate (100X Concentrate)
Click to view available options
Quantity:
2mL
4mL
10mL
Unit Size:
Each
Description
The Galacton-Plus™ Substrate is a concentrated chemiluminescent substrate for E.coli β-galactosidase assays.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Specifications
| Content And Storage | Store the product at 4°C. |
| Form | Liquid |
| Product Line | Galacton-Plus™ |
| Quantity | 4mL |
| Substrate | Beta-Galactosidase |
| Substrate Type | Beta-Gal Substrate |
| Format | Bottle(s) |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction