missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Galacton-Plus™ Substrate (100X Concentrate)
Galacton-Plus™ Substrate (100X Concentrate)
Brand: Molecular Probes™ T2120
Code : NEW
Additional Details : Weight : 0.02000kg
Description
The Galacton-Plus™ Substrate is a concentrated chemiluminescent substrate for E.coli β-galactosidase assays.For Research Use Only. Not for use in diagnostics procedures.
Specifications
Liquid | |
Room Temperature | |
Beta-Gal Substrate | |
10mL |
Galacton-Plus™ | |
Beta-Galactosidase | |
Bottle(s) | |
Store the product at 4°C. |
For Research Use Only. Not for use in diagnostic procedures.