missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gal-Screen™ β-Galactosidase Reporter Gene Assay System for Mammalian Cells
Combines direct cell lysis with rapid, ultra-sensitive chemiluminescent detection of β-galactosidase reporter enzyme.
£217.00 - £4515.00
Specifications
Product Type | β-Galactosidase Reporter Gene Assay System |
---|---|
Detection Method | Chemiluminescent |
For Use With (Equipment) | Luminometer (Microplate) |
Assay | β-Gal (lacZ) Assay |
Compatible Cells | Mammalian Cells |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12654285
|
Invitrogen™
T1027 |
1000 Assays |
£748.00
Each |
Estimated Shipment: 10-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12664285
|
Invitrogen™
T1028 |
10,000 Assays |
£4515.00
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12674285
|
Invitrogen™
T1029 |
200 Assays |
£217.00
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Homogeneous assay format allows detection of β-galactosidase in presence of normal culture media without removal of media and without an additional cell lysis step, providing the easiest and most streamlined assay procedure possible
- The kinetics of glow-assay provides a “window” during which measurements may be performed, facilitating HTS applications where assay automation is used
- Can be used with either mammalian or yeast model systems, providing flexibility in choice of model systems
- Wide dynamic range of β-galactosidase assay enables accurate measurement of enzyme from femtogram to nanogram range
- Assay sensitivity is 100 to 1,000-fold better than either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assay technologies
- Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions
- Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes
- Assay can be completed in about one hour, providing fast assay turnaround
Ideal for Screening
This homogeneous assay is ideally suited for screening applications where assay automation is required. The Gal-Screen system uses Galacton-Star™ chemiluminescent substrate for convenient measurement in a luminometer. Light emission reaches maximum in 60-90 minutes and remains constant for 45-90 minutes.
Specifications
- Assay Type: β-Gal (lacZ) Assay
- Compatible Cells: Mammalian Cells
- Detection Method: Chemiluminescent
- For Use With Equipment: Luminometer (Microplate)
- Form: Liquid
- Format: 96-well plate
- Readout: End Point
- Sample Type (Specific): Cell Cultures
- Substrate: Galacton-Star™
- Substrate Properties: Chemical Substrate
- Substrate Type: Beta-Gal Substrate
- Target Enzyme: Beta-Galactosidase
- Technique: Enhanced Chemiluminescence
Specifications
β-Galactosidase Reporter Gene Assay System | |
Luminometer (Microplate) | |
Mammalian Cells | |
Unconjugated | |
Cell Cultures | |
Beta-Gal Substrate | |
Beta-Galactosidase | |
Enzyme Assay |
Chemiluminescent | |
β-Gal (lacZ) Assay | |
High-throughput Compatible | |
End Point | |
Galacton-Star™ | |
Chemical Substrate | |
Gal-Screen™, NovaBright™ | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.