Learn More
Alfa Aesar™ Gadolinium sputtering target, 50.8mm dia x 1.59mm thick
Brand: Alfa Aesar™ 045621.KS
Code : NEW
Additional Details : CAS Number : 7440-54-2 Weight : 0.01000kg
Quantity | 1 ea. |
---|
Specifications
Gadolinium sputtering target | |
50.8mm dia. x 1.59mm thick | |
MFCD00011022 |
1 ea. | |
7440-54-2 |