missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium rod, 19mm (0.75 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific™

Catalog Number p-7058687
Click to view available options
Quantity:
10 mm
25 mm
This item is not returnable. View return policy
7440-54-2
Gd
157.25
MFCD00011022
UIWYJDYFSGRHKR-UHFFFAOYSA-N
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
23982
CHEBI:33375
gadolinium
[Gd]
This item is not returnable. View return policy
7440-54-2
Gd
157.25
MFCD00011022
UIWYJDYFSGRHKR-UHFFFAOYSA-N
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
23982
CHEBI:33375
gadolinium
[Gd]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged in mylar under argon
TRUSTED_SUSTAINABILITY
Quantity 10 mm
Percent Purity 99.9%
Packaging Mylar under Argon
Weight ≈2.2g/mm
Form Cast Rod
Assay (metals basis excluding Ta)
Chemical Name or Material Gadolinium rod, ≈19mm (0.75 in.) dia.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'