missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadolinium rod, 19mm (0.75 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific™
Click to view available options
Quantity:
10 mm
25 mm
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged in mylar under argon
Specifications
Quantity | 10 mm |
Percent Purity | 99.9% |
Packaging | Mylar under Argon |
Weight | ≈2.2g/mm |
Form | Cast Rod |
Assay | (metals basis excluding Ta) |
Chemical Name or Material | Gadolinium rod, ≈19mm (0.75 in.) dia. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'