missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) isopropoxide, 99.9% (metals basis), 5% w/v in toluene/isopropanol (3:2), Thermo Scientific Chemicals

Catalog Number p-7051931
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
14532-05-9
C9H21GdO3
334.51
MFCD00144381
VJLSFXQJAXVOEQ-UHFFFAOYSA-N
gadolinium iii isopropoxide, triisopropoxygadolinium, gadolinium isopropoxide, acmc-20akue, gadolinium triisopropoxide, gadolinium tripropan-2-olate, gadolinium-i-propoxide, gadolinium 3+ ion tris propan-2-olate, gadolinium 3+ tris propan-2-olate
15047187
gadolinium(3+);propan-2-olate
[Gd+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
This item is not returnable. View return policy
14532-05-9
C9H21GdO3
334.51
MFCD00144381
VJLSFXQJAXVOEQ-UHFFFAOYSA-N
gadolinium iii isopropoxide, triisopropoxygadolinium, gadolinium isopropoxide, acmc-20akue, gadolinium triisopropoxide, gadolinium tripropan-2-olate, gadolinium-i-propoxide, gadolinium 3+ ion tris propan-2-olate, gadolinium 3+ tris propan-2-olate
15047187
gadolinium(3+);propan-2-olate
[Gd+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]

Precursor for Gd2O3 films using Sol-Gel process

It is employed in the catalyst action which is used in a ring opening of meso-aziridines with trimethylsilyl azide. In many asymmetric catalysis applications, glove box and Schlenk techniques should be employed to prevent exposure of the rare earth catalyst to air and moisture, which can be detrimental to the reaction outcome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Odor Aromatic
Linear Formula Gd[OCH(CH3)2]3
Quantity 25 mL
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes slowly in water.
Formula Weight 334.51
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 5% w/v in toluene/isopropanol (3:2)
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Gadolinium(III) isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'