missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gadolinium(III) isopropoxide, 99.9% (metals basis), 5% w/v in toluene/isopropanol (3:2)

Precursor for Gd2O3 films using Sol-Gel process

Brand:  Alfa Aesar™ 042277.AE

Code : NEW

Additional Details : CAS Number : 14532-05-9 Weight : 0.01000kg Transport : UN number : 1993 Chem class : 3 Pack group : II

Quantity 100mL

 View more versions of this product

Product Code. 11319967

  • £673.00 / 100mL


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

Gadolinium(III) isopropoxide
Gd[OCH(CH3)2]3
UN1993
14532-05-9
MFCD00144381

Specifications

Gadolinium(III) isopropoxide
Moisture Sensitive
14532-05-9
99.9%
Gd[OCH(CH3)2]3
UN1993
Pale yellow
5% w/v in toluene/isopropanol (3:2)
100mL
Liquid
(metals basis)
MFCD00144381
Aromatic
Videos
SDS