missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11314118
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13765-26-9
F3Gd
214.25
MFCD00016113
TYIZUJNEZNBXRS-UHFFFAOYSA-K
gadolinium(3+) trifluoride
[F-].[F-].[F-].[Gd+3]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13765-26-9
F3Gd
214.25
MFCD00016113
TYIZUJNEZNBXRS-UHFFFAOYSA-K
gadolinium(3+) trifluoride
[F-].[F-].[F-].[Gd+3]

For vacuum deposition

Gadolinium(III) fluoride, anhydrous is employed in the synthesis of non-oxide glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1231°C
Color White
Physical Form Pieces
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 214.25
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 7.1 g/mL
Assay (REO)
Chemical Name or Material Gadolinium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'