missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7050156
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
13818-75-2
MFCD00049741
This item is not returnable. View return policy
13818-75-2
MFCD00049741

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point 770°C
Physical Form Powder
Quantity 2 g
Formula Weight 396.96
Percent Purity 99.9%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Gadolinium(III) bromide, Ultra dry

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'