Learn More
Thermo Scientific Alfa Aesar Gadolinium gallium isopropoxide, 99.9% (metals basis), 25% w/v in isopropanol
Brand: Thermo Scientific Alfa Aesar 036582.AB
Gadolinium gallium isopropoxide, 99.9% (metals basis), 25% w/v in isopropanol
Code : NEW
Additional Details : CAS Number : 51062-97-6 Weight : 0.01000kg Transport : UN number : 1219 Chem class : 3 Pack group : II