missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Gadolinium, AAS standard solution, Specpure™, Gd 1000μg/mL

Brand:  Alfa Aesar™ 088065.AP

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 500mL

 View more versions of this product

Product Code. 11316349

  • POA / 500mL

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

Gadolinium, AAS standard solution
Specpure™
UN3264
Gd 1000μg/mL
MFCD00011022

Specifications

Gadolinium, AAS standard solution
Specpure™
Gd2O3 in 5% HNO3
UN3264
Gd 1000μg/mL
Liquid
MFCD00011022
500mL
Videos
SDS