missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ GAD2 (GAD65) Recombinant Mouse Protein, N-His.GST Tag

Expressed from DNA sequence encoding full length of mouse GAD2 (Met 1-Leu 585) fused to N-terminal polyhistidine-tagged GST tag at N-terminus

Brand:  Sino Biological™ 50653M20B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15339106

  • £369.00 / 50µg

Estimated Shipment date
from Supplier 03-06-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM PMSF, 0.5mM GSH, 3mM DTT, 10% glycerol, pH 8.0
Specifications

Specifications

Insect
Assay Standard
Lyophilized
93kd
50μg
Full Length, Recombinant
Glutamate Decarboxylase 2 (pancreatic islets and brain 65kDa)
GAD2 (GAD65) Recombinant Mouse Protein
Other Proteins
Analysis of protein stucture and used in cell biology research applications
14417
≥90% by SDS-PAGE
Mouse
<1 EU/μg
GAD2
GST Tag, His Tag
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.