Learn More
Gabexate mesylate, Tocris Bioscience™
Serine protease inhibitor; inhibits thrombin, trypsin, kallikrein and plasmin
Brand:  Tocris (Bio-Techne) 179810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13440024
  • £144.06 / 10mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Serine protease inhibitor; inhibits trypsin, plasmin, plasma kallikrein and thrombin (IC50 values are 9.4, 30, 41 and 110 μM respectively). Antithrombotic in vitro and in vivo. Also inhibits LPS-induced TNF-α production, probably by inhibiting NF-κB and AP-1 activation.
Specifications
Chemical Identifiers
56974-61-9
417.477
gabexate mesylate, gabexate mesilate, gabexate monomethanesulfonate, megacart, gabexate methanesulfonate, foy, dsstox_cid_25591, dsstox_rid_80986, dsstox_gsid_45591
ethyl 4-[6-(diaminomethylideneamino)hexanoyloxy]benzoate;methanesulfonic acid
C17H27N3O7S
DNTNDFLIKUKKOC-UHFFFAOYSA-N
6604561
CCOC(=O)C1=CC=C(C=C1)OC(=O)CCCCCN=C(N)N.CS(=O)(=O)O
Specifications
10mg
56974-61-9
gabexate mesylate, gabexate mesilate, gabexate monomethanesulfonate, megacart, gabexate methanesulfonate, foy, dsstox_cid_25591, dsstox_rid_80986, dsstox_gsid_45591
CCOC(=O)C1=CC=C(C=C1)OC(=O)CCCCCN=C(N)N.CS(=O)(=O)O
417.477
417.48
Gabexate mesylate
C17H27N3O7S
DNTNDFLIKUKKOC-UHFFFAOYSA-N
ethyl 4-[6-(diaminomethylideneamino)hexanoyloxy]benzoate;methanesulfonic acid
6604561
>98%