missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals G3335
Click to view available options
Quantity:
50 mg
100 mg
Description
A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist
Specifications
Quantity | 100 mg |
Description | White |
Content And Storage | 2°C to 8°C |
For Use With (Application) | A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist |
Formula Weight | 333.34g/mol |
Sensitivity | Light Sensitive |
Product Type | Signal Transduction Reagent |
Form | Solid |
InChI Key | PWIQCLSQVQBOQV-UHFFFAOYNA-N |
SMILES | NC(CC1=CNC2=CC=CC=C12)C(=O)NC(CCC(O)=O)C(O)=O |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'