Learn More
G-CSF Monoclonal Antibody (9B4CSF), eFluor™ 660, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£82.75 - £249.00
Specifications
Antigen | G-CSF |
---|---|
Clone | 9B4CSF |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15550037
|
Affymetrix eBioscience
50-7353-82 |
100 μg |
£249.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15540037
|
Affymetrix eBioscience
50-7353-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This 9B4CSF monoclonal antibody reacts with mouse G-CSF (Granulocyte Colony-Stimulating Factor), a growth factor that stimulates the proliferation, differentiation, and mobilization of hematopoetic stem cells. It is expressed by monocytes, macrophages, and bone marrow stromal cells, and can also be induced in fibroblasts by IL-17A. Unlike GM-CSF and IL-3, which can stimulate cells of multiple lineages, G-CSF activity is limited to neutrophilic granulocytes. G-CSF is essential to the maintece of neutrophil counts during homeostasis, and low basal levels of the protein are detectable in the serum of healthy individuals. Circulating levels become elevated rapidly upon infection, as G-CSF is also important for the activation and mobilization of mature neutrophils during the innate immune response. G-CSF therapy is commonly used to treat neutropenia following bone marrow transplant or chemotherapy. This 9B4CSF antibody has been demonstrated to cross-react with human G-CSF. Applications Reported: This 9B4CSF antibody has been reported for use in flow cytometric analysis, and intracellular staining followed by flow cytometric analysis. Applications Tested: This 9B2-8°CSF antibody has been tested by intracellular staining followed by flow cytometric analysis of in vitro-cultured and stimulated mouse monocytes/macrophages. This can be used at less than or equal to 0.06 μg per test.
G-CSF Monoclonal antibody specifically detects G-CSF in Mouse samples. It is validated for Flow Cytometry.Specifications
G-CSF | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P09920 | |
Csf3 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Csf3 |
9B4CSF | |
Flow Cytometry | |
eFluor 660 | |
Rat | |
Mouse | |
12985 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.