missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Furin Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
POA
Specifications
Antigen | Furin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Furin Polyclonal specifically detects Furin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Furin | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FUR, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACE, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PC, PCSK3, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1 | |
FURIN | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
P09958 | |
5045 | |
Mouse myeloma cell line NS0-derived recombinant human Furin Asp108-Glu715 Accession # P09958 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Furin in direct ELISAs and Western blots. In direct ELISAs, less than 40% cross-reactivity with recombinant mouse Furin is observed, and less than 1% cross-reactivity with recombinant human (rh) PC-1 and rhPC-2 is observed. |
For Research Use Only