missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl glycidyl ether, 97%, Thermo Scientific Chemicals

Catalog Number p-137528
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5380-87-0
C8H10O3
154.17
MFCD00013348
RUGWIVARLJMKDM-UHFFFAOYNA-N
furfuryl glycidyl ether, 2-oxiran-2-ylmethoxy methyl furan, 2,3-epoxypropyl-furfurylether, 2-oxiranylmethoxy methyl furan, 2-2,3-epoxypropoxy methyl furan, furan, 2-oxiranylmethoxy methyl, furan, 2-2,3-epoxypropoxy methyl
94306
2-(oxiran-2-ylmethoxymethyl)furan
C(OCC1=CC=CO1)C1CO1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5380-87-0
C8H10O3
154.17
MFCD00013348
RUGWIVARLJMKDM-UHFFFAOYNA-N
furfuryl glycidyl ether, 2-oxiran-2-ylmethoxy methyl furan, 2,3-epoxypropyl-furfurylether, 2-oxiranylmethoxy methyl furan, 2-2,3-epoxypropoxy methyl furan, furan, 2-oxiranylmethoxy methyl, furan, 2-2,3-epoxypropoxy methyl
94306
2-(oxiran-2-ylmethoxymethyl)furan
C(OCC1=CC=CO1)C1CO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Orange
Density 1.1220g/mL
Boiling Point 103.0°C to 104.0°C (11.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4800 to 1.4820
Quantity 5 g
Specific Gravity 1.122
Formula Weight 154.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Furfuryl glycidyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'