missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furan, 99+%, stabilized, Thermo Scientific Chemicals
16.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -85.6°C |
Color | Colorless to Yellow |
Density | 0.9360g/mL |
Boiling Point | 31.3°C |
Flash Point | −35°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 0.05% max. (Furfural) (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4197 to 1.4217 |
Quantity | 250 mL |
Show More |
Safety and Handling
Product Identifier
- Furan, stabilized
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Flammable liquid Category 1
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H350-May cause cancer.
- H315-Causes skin irritation.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H224-Extremely flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'