missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Fura-2 Calcium Imaging Calibration Kit (Zero to 10 mM CaEGTA, 50 μM Fura-2)
Fura-2 Calcium Imaging Calibration Kit (Zero to 10 mM CaEGTA, 50 μM Fura-2)
Brand: Molecular Probes™ F6774
214.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.09000kg
Description
The Fura-2 Calcium Imaging Calibration Kit is designed to facilitate rapid calibration and standardization of digital imaging microscopes. The kit contains 11 pre-diluted buffers supplemented with 50μM fura-2 (F-1200) as well as 15μm—diameter microspheres.Specifications
Store in refrigerator 2°C to 8°C and protect from light. | |
Calcium Imaging Calibration | |
Reagent | |
Room Temperature |
Fluorescent | |
Fluorescence Microscope | |
Ion Calibration Reagent |
For Research Use Only. Not for use in diagnostic procedures.