missing translation for 'onlineSavingsMsg'
Learn More

β-Funaltrexamine hydrochloride, Tocris Bioscience™

Irreversible and selective μ antagonist

Brand:  Tocris (Bio-Techne) 0926/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13428163

  • £208.74 / 10mg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective μ opioid receptor antagonist.
Specifications

Chemical Identifiers

72786-10-8
490.981
(E)-4-[[5α,6β)-17-Cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxymorphinan-6-yl]amino]-4-oxo-2-butenoicacid methyl ester hydrochloride
methyl (E)-4-[[(4R,4aR,7R,7aR,12bR)-3-(cyclopropylmethyl)-4a,9-dihydroxy-1,2,4,5,6,7,7a,13-octahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-yl]amino]-4-oxobut-2-enoate;hydrochloride
C25H31ClN2O6
BIPHUOBUKMPSQR-RMEGLGEHSA-N
124081002
COC(=O)C=CC(=O)NC1CCC2(C3CC4=C5C2(C1OC5=C(C=C4)O)CCN3CC6CC6)O.Cl

Specifications

10mg
72786-10-8
(E)-4-[[5α,6β)-17-Cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxymorphinan-6-yl]amino]-4-oxo-2-butenoicacid methyl ester hydrochloride
COC(=O)C=CC(=O)NC1CCC2(C3CC4=C5C2(C1OC5=C(C=C4)O)CCN3CC6CC6)O.Cl
490.981
490.99
β-Funaltrexamine hydrochloride
C25H31ClN2O6
BIPHUOBUKMPSQR-RMEGLGEHSA-N
methyl (E)-4-[[(4R,4aR,7R,7aR,12bR)-3-(cyclopropylmethyl)-4a,9-dihydroxy-1,2,4,5,6,7,7a,13-octahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-yl]amino]-4-oxobut-2-enoate;hydrochloride
124081002
>98%
Videos
Documents