missing translation for 'onlineSavingsMsg'
Learn More

Labcaire™ Fume cabinet 750E 2 x main filter only 230V 50Hz 820W Aura

Brand:  Labcaire™ AU-750E

Code : 04

Additional Details : Weight : 151.00000kg

Product Code. 12801800

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.