missing translation for 'onlineSavingsMsg'
Learn More

Fumaraldehyde bis(dimethylacetal), 90%, Thermo Scientific™

Catalog Number p-3523455
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6068-62-8
C8H16O4
176.21
MFCD00075507
ZFGVCDSFRAMNMT-AATRIKPKSA-N
fumaraldehyde bis dimethyl acetal, e-1,1,4,4-tetramethoxybut-2-ene, fumaraldehyde bis dimethylacetal, 2-butenedial bis dimethyl acetal, 1,1,4,4-tetramethoxy-trans-2-butene, trans-1,1,4,4-tetramethoxy-2-butene, 1,1,4,4-tetramethoxy-but-2-ene
5862719
(E)-1,1,4,4-tetramethoxybut-2-ene
COC(C=CC(OC)OC)OC
This item is not returnable. View return policy
6068-62-8
C8H16O4
176.21
MFCD00075507
ZFGVCDSFRAMNMT-AATRIKPKSA-N
fumaraldehyde bis dimethyl acetal, e-1,1,4,4-tetramethoxybut-2-ene, fumaraldehyde bis dimethylacetal, 2-butenedial bis dimethyl acetal, 1,1,4,4-tetramethoxy-trans-2-butene, trans-1,1,4,4-tetramethoxy-2-butene, 1,1,4,4-tetramethoxy-but-2-ene
5862719
(E)-1,1,4,4-tetramethoxybut-2-ene
COC(C=CC(OC)OC)OC
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 100°C to 103°C (15.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2CHCH=CHCH(OCH3)2
Quantity 1 g
Specific Gravity 1.01
Formula Weight 176.21
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Fumaraldehyde bis(dimethylacetal)
Show More Show Less
compliance-icons
Product Identifier
  • Fumaraldehyde bis(dimethylacetal)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'