Learn More
Thermo Scientific Alfa Aesar Fullerene powder, mixed refined, typically 77% C{60}, 22% C{70}, <2% higher
Brand: Thermo Scientific Alfa Aesar 040968.03
Fullerene powder, mixed refined, typically 77% C{60}, 22% C{70}, <2% higher
Code : NEW
Additional Details : CAS Number : 99685-96-8 Weight : 0.00100kg Transport : UN number : 1325 Chem class : 4.1 Pack group : III