missing translation for 'onlineSavingsMsg'
Learn More

Fullerene powder, mixed refined, typically 70% C{60}, 28% C{70}, higher 2%, Thermo Scientific Chemicals

Catalog Number 11309557
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
99685-96-8
C60
720.66
MFCD00282904
XMWRBQBLMFGWIX-UHFFFAOYSA-N
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
123591
CHEBI:33128
(C\{60}-I\{h})[5,6]fullerene
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
This item is not returnable. View return policy
99685-96-8
C60
720.66
MFCD00282904
XMWRBQBLMFGWIX-UHFFFAOYSA-N
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
123591
CHEBI:33128
(C\{60}-I\{h})[5,6]fullerene
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black-Brown
Quantity 250 mg
UN Number UN1325
Concentration or Composition (by Analyte or Components) typically 73% C60, 22% C70, higher 5%
Physical Form Powder
Chemical Name or Material Fullerene powder, Mixed refined

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'