missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Fullerene powder, mixed hydrogenated, typically 77% C{60}H{x}, 22% C{70}H{y}

Brand:  Alfa Aesar™ 040967.77

Code : NEW

Additional Details : Weight : 0.00010kg Transport : UN number : 1325 Chem class : 1 Pack group : III

Quantity 0.1g

 View more versions of this product

Product Code. 11315297

  • £63.18 / 0.10g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

Fullerene powder
MFCD00282904
C70
Mixed hydrogenated
UN1325

Specifications

Fullerene powder
Powder
UN1325
0.1g
Mixed hydrogenated
MFCD00282904
C70
typically 77% C60Hx, 22% C70Hy
Videos
SDS