missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fullerene powder, 98+% C{70}, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Fullerene powder is used in chemical research. It is extensively used for several biomedical applications including the design of high-performance MRI contrast agents, X-Ray imaging contrast agents, photodynamic therapy and drug and gene delivery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black |
Density | ≈1.7 g/mL |
Boiling Point | 500°C to 600°C (sublimation) |
Quantity | 250 mg |
UN Number | UN1325 |
Solubility Information | Soluble in toluene and benzene. |
Percent Purity | ≥98% |
Physical Form | Powder |
Assay | C70 |
Chemical Name or Material | Fullerene powder |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'