missing translation for 'onlineSavingsMsg'
Learn More

Fullerene powder, 97% C{70}, Thermo Scientific Chemicals

Catalog Number p-7050920
Click to view available options
Quantity:
10 mg
50 mg
250 mg
This item is not returnable. View return policy
115383-22-7
C70
840.77
MFCD00282904
ATLMFJTZZPOKLC-UHFFFAOYSA-N
fullerene, fullerene 70, rugbyballene, carbon, 70-d5h fullerene, buckminsterfullerene, 5,6 fullerene-c70-d5h 6, c70-d5h 6 5,6 fullerene, fullerene powder, c 250mg
16131935
CHEBI:33195
(C\{70}-D\{5h(6)})[5,6]fullerene
C12=C3C4=C5C6=C7C8=C9C%10=C%11C%12=C%13C%10=C%10C8=C5C1=C%10C1=C%13C5=C8C1=C2C1=C3C2=C3C%10=C%13C%14=C3C1=C8C1=C3C5=C%12C5=C8C%11=C%11C9=C7C7=C9C6=C4C2=C2C%10=C4C(=C29)C2=C6C(=C8C8=C9C6=C4C%13=C9C(=C%141)C3=C85)C%11=C27
This item is not returnable. View return policy
115383-22-7
C70
840.77
MFCD00282904
ATLMFJTZZPOKLC-UHFFFAOYSA-N
fullerene, fullerene 70, rugbyballene, carbon, 70-d5h fullerene, buckminsterfullerene, 5,6 fullerene-c70-d5h 6, c70-d5h 6 5,6 fullerene, fullerene powder, c 250mg
16131935
CHEBI:33195
(C\{70}-D\{5h(6)})[5,6]fullerene
C12=C3C4=C5C6=C7C8=C9C%10=C%11C%12=C%13C%10=C%10C8=C5C1=C%10C1=C%13C5=C8C1=C2C1=C3C2=C3C%10=C%13C%14=C3C1=C8C1=C3C5=C%12C5=C8C%11=C%11C9=C7C7=C9C6=C4C2=C2C%10=C4C(=C29)C2=C6C(=C8C8=C9C6=C4C%13=C9C(=C%141)C3=C85)C%11=C27

Fullerene powder is used in chemical research. It is extensively used for several biomedical applications including the design of high-performance MRI contrast agents, X-Ray imaging contrast agents, photodynamic therapy and drug and gene delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black
Density 1.7 g/mL
Boiling Point 500°C to 600°C (sublimation)
Quantity 10 mg
UN Number UN1325
Solubility Information Soluble in toluene and benzene.
Percent Purity 97%
Physical Form Powder
Assay C70
Chemical Name or Material Fullerene powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'