missing translation for 'onlineSavingsMsg'
Learn More

Fullerene, nanotube, multi-walled, 20 nm OD, 5-20 micron long, Thermo Scientific Chemicals

Catalog Number p-7059037
Click to view available options
Quantity:
50 mg
250 mg
This item is not returnable. View return policy
308068-56-6
CH4
16.043
MFCD00133992
VNWKTOKETHGBQD-UHFFFAOYSA-N
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
297
CHEBI:16183
methane
C
This item is not returnable. View return policy
308068-56-6
CH4
16.043
MFCD00133992
VNWKTOKETHGBQD-UHFFFAOYSA-N
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
297
CHEBI:16183
methane
C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black
Mesh Size −270 Mesh
Quantity 50 mg
UN Number UN1325
Physical Form Powder
Chemical Name or Material Fullerene, Nanotube, multi-walled, 20nm OD, 5 to 20 micron long

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'