Learn More
Fullerene, nanotube, multi-walled, 20-50 nm OD, 5-20 micron long, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Carbon nanotubes enhance the electrical conductivity and the mechanical properties. It is also used in chemical research. It is suitable as a hydrogen storage material. It is an ideal super capacitor electrode materials, It is used in the preparation of super composites. carbon nanotubes have excellent field emission performance and used in heavy weight cathode tube technology. It is also applied to naanometer electronic, catalytic fiber machine. Nanotubes will have an important role in nanotechnology engineering. carbon nanotube also can be used in making molecule bearings and nano robot.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black |
Quantity | 5 g |
UN Number | UN1325 |
Solubility Information | It is miscible,can be diluted with water. |
Physical Form | Powder |
Chemical Name or Material | Fullerene, Nanotube, multi-walled, 20 to 50nm OD, 5 to 20 micron long |
RUO – Research Use Only