missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Fullerene, nanotube, multi-walled, 20-50 nm OD, <1 micron long

Brand:  Alfa Aesar™ 044197.06

Code : NEW

Additional Details : CAS Number : 308068-56-6 Weight : 0.00500kg Transport : UN number : 1325 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15486585

  • £954.64 / 5g


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

Fullerene
Nanotube, multi-walled, 20 to 50nm OD, <1 micron long
UN1325
308068-56-6
MFCD00133992

Specifications

Fullerene
308068-56-6
MFCD00133992
Black
Nanotube, multi-walled, 20 to 50nm OD, <1 micron long
Powder
UN1325
5g
Videos
SDS