missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Fullerene, carbon nanotube, multi-walled, 3-20 nm OD, 1-3 nm ID, 0.1-10 micron long, 95%

Brand:  Alfa Aesar™ 044945.06

Code : NEW

Additional Details : CAS Number : 308068-56-6 Weight : 0.00500kg Transport : UN number : 1325 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15444925

  • £123.70 / 5g


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Specifications

Chemical Identifiers

Fullerene, carbon nanotube
Multi-walled, OD: 3 to 20nm, ID: 1 to 3nm, 0.1 to 10 micron long
UN1325
308068-56-6
MFCD06202029

Specifications

Fullerene, carbon nanotube
308068-56-6
95%
UN1325
5g
Multi-walled, OD: 3 to 20nm, ID: 1 to 3nm, 0.1 to 10 micron long
Powder
MFCD06202029
Black
Videos
SDS