missing translation for 'onlineSavingsMsg'
Learn More

Fullerene, buckytube/nanotube, single walled, Thermo Scientific Chemicals

Catalog Number p-7059394
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
308068-56-6
CH4
16.043
MFCD00282904
VNWKTOKETHGBQD-UHFFFAOYSA-N
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
297
CHEBI:16183
methane
C
This item is not returnable. View return policy
308068-56-6
CH4
16.043
MFCD00282904
VNWKTOKETHGBQD-UHFFFAOYSA-N
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
297
CHEBI:16183
methane
C

Carbon nanotubes enhance the electrical conductivity and the mechanical properties. It is also used in chemical research. It is suitable as a hydrogen storage material. It is an ideal super capacitor electrode materials, It is used in the preparation of super composites. carbon nanotubes have excellent field emission performance and used in heavy weight cathode tube technology. It is also applied to naanometer electronic, catalytic fiber machine. Nanotubes will have an important role in nanotechnology engineering. carbon nanotube also can be used in making molecule bearings and nano robot.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black
Quantity 0.25 g
UN Number UN1325
Solubility Information It is miscible,can be diluted with water. Soluble in benzene,toluene.
Physical Form Powder
Chemical Name or Material Fullerene, Buckytube/nanotube, single walled

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'