missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Fullerene, buckytube/nanotube, multi-walled, ground core, 7-12nm OD, 0.5-10 micron long

Brand:  Alfa Aesar™ 041549.14

Code : NEW

Additional Details : CAS Number : 308068-56-6 Weight : 0.02500kg Transport : UN number : 1325 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11364308

  • £621.00 / 25g


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

Fullerene, buckytube/nanotube
Multi-walled, ground core, 7 to 12nm OD, 0.5 to 10 micron long
UN1325
308068-56-6
MFCD00282904

Specifications

Fullerene, buckytube/nanotube
308068-56-6
MFCD00282904
Black
−100 Mesh
Multi-walled, ground core, 7 to 12nm OD, 0.5 to 10 micron long
Powder
UN1325
25g
Videos
SDS