missing translation for 'onlineSavingsMsg'
Learn More

Fullerene, buckytube/nanotube, double-walled, 50 - 80%, Thermo Scientific™

Catalog Number p-7059485
Click to view available options
Quantity:
0.1 g
0.5 g
2 g
This item is not returnable. View return policy
308068-56-6
CH4
16.043
MFCD00282904
VNWKTOKETHGBQD-UHFFFAOYSA-N
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
297
CHEBI:16183
methane
C
This item is not returnable. View return policy
308068-56-6
CH4
16.043
MFCD00282904
VNWKTOKETHGBQD-UHFFFAOYSA-N
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
297
CHEBI:16183
methane
C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 0.1 g
UN Number UN1325
Percent Purity 50 to 80%
Physical Form Powder
Chemical Name or Material Fullerene, Buckytube/nanotube, double-walled
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'