missing translation for 'onlineSavingsMsg'
Learn More

Ftorafur, 99%, ACROS Organics™

Brand:  Acros Organics 197702500

Code : 29

Additional Details : CAS Number : 17902-23-7 Weight : 0.05025kg


Quantity 250mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10712821

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Ftorafur
17902-23-7
99%
200.17
5-Fluoro-1-(tetrahydro-2-furyl)uracil
99%
99%
C8H9FN2O3
MFCD00012351

Specifications

Ftorafur
17902-23-7
Very Fine Crystalline Powder
99%
C8H9FN2O3
MFCD00012351
5-Fluoro-1-(tetrahydro-2-furyl)uracil
168.0° to 171.0°C
99%
Authentic
99%
Glass bottle
200.17
15, 9252
White
250mg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ftorafur

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS