missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ FTO Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding human FTO (Lys 2-Pro 505) fused to polyhistide tag at N-terminus

Brand:  Sino Biological™ 12125H07E50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15285775

  • £370.60 / 50µg


Estimated Shipment
06-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: 20mM Tris, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween80, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
79608
≥90% by SDS-PAGE
Human
Fat mass and Obesity associated
Assay Standard
E.coli
His Tag
Lyophilized
59.5kd
50μg
Full Length, Recombinant
FTO
Methylation and Demethylation Proteins
FTO Recombinant Human Protein
Videos