missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ FTO Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding human FTO (Lys 2-Pro 505) fused to polyhistide tag at N-terminus

Brand:  Sino Biological™ 12125H07E5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15275775

  • £91.09 / 5µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

  • Formulation: 20mM Tris, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween80, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
79608
≥90% by SDS-PAGE
Human
Fat mass and Obesity associated
Assay Standard
E.coli
His Tag
Lyophilized
59.5kd
5μg
Full Length, Recombinant
FTO
Methylation and Demethylation Proteins
FTO Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.