missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ FTL Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding human FTL (Met 1-Asp 175) fused to polyhistidine tag at N-terminus

Brand:  Sino Biological™ 12482H07E50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15235955

  • £158.24 / 50µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: 50mM Tris, 20% glycerol, pH 9.5
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
2512
≥90% by SDS-PAGE
Human
Ferritin Light polypeptide
Assay Standard
E.coli
N-His Tag
Lyophilized
22.1kd
50μg
Full Length, Recombinant
FTL
Other Proteins
FTL Recombinant Human Protein
Videos