Learn More
FRY Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559655
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EDVLFGFTNF LLREVNDMHH TLLDSSLKLL LQLLTQWKLV IQTQGKVYEQ ANKIRNSELI ANGSSHRIQS ERGPHCSVLH AVE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 87%.
FRY Polyclonal antibody specifically detects FRY in Human samples. It is validated for Immunohistochemistry.Specifications
FRY | |
Polyclonal | |
Unconjugated | |
FRY | |
13CDNA73; 214K23.2; bA207N4.2; bA37E23.1; C13orf14; CG003; FRY; FRY microtubule binding protein; furry homolog; protein furry homolog; WUGSC:H_2G3A.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10129 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TBA9 | |
FRY | |
Recombinant protein corresponding to Human FRY | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only