missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRK (PTK5) Recombinant Human Protein
FRK (PTK5) is a nuclear protein.
£1602.00 - £6900.00
Specifications
Accession Number | NP_002022 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 86.3kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488613
|
Invitrogen™
PR7729A |
100μg |
£1602.00
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10056892
|
Invitrogen™
PV4192 |
1mg |
£6900.00
1mg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FRK (PTK5) may function during G1 and S phase of the cell cycle, suppressing growth.
Specifications
NP_002022 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
SRC Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
86.3kDa | |
RUO - research use only | |
FRK (PTK5), GTK, RAK | |
Human | |
GST Tag | |
Frozen |