Freudenberg™ Roll-O-Matic™ Self-Wringing Mops Refills

Suitable for all controlled environments

Overview
Brand: Vileda

Manufacturer Part Number: 119656

UNSPSC: 47131600

Code: NEW

Additional Details:
Additional Details: Weight: 2.50000kg



Product Code. 15203556

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

Product Type Roller Mop Refill
Color Blue
Material Polyester with stainless-steel channel
For Use With (Application) For tear free application of disinfectants, cleaning and spill absorption
For Use With (Equipment) Original and self wringing roll-O-matic roller mops