Freudenberg™ Roll-O-Matic™ Self-Wringing Mops Refills

Suitable for all controlled environments

Overview
Brand: Vileda

Manufacturer Part Number: 119616

X12 SPONGE MOP REFILL WITH PES LAMINATION 10''

UNSPSC: 47131600

Code: NEW

Additional Details:
Additional Details: Weight: 2.00000kg



Product Code. 15293546

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

Color Blue
For Use With (Application) For tear free cleaning of deep contaminants and disinfectant residues
For Use With (Equipment) Original and self wringing roll-O-matic roller mops
Autoclavable Yes
Material Polyester, Stainless Steel
Product Type Refill