Vileda Professional™ Roll-O-Matic™ Self-Wringing Mops

Suitable for all controlled environments.

Overview
Brand: Vileda

Manufacturer Part Number: 118550

ROLL-O-MATIC MOP HANDLE STAINLESS - 14'' REFILL

UNSPSC: 47131600

Code: NEW

Additional Details:
Additional Details: Weight: 2.00000kg



Product Code. 15243546

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

Autoclavable Yes
Color Blue
For Use With (Application) For use on floors, walls, and ceilings in cleanroom environments. Ideal for the pull-and-lift cleaning technique
For Use With (Equipment) Roll-O-matic self wringing cleanroom mops
Includes Wringer built into handle
Material Stainless Steel
Product Type Stainless Steel Mops with Sponge Mop Head, Straight Headl