Vileda Professional™ Roll-O-Matic™ Self-Wringing Mops

Suitable for all controlled environments.

Overview
Brand: Vileda

Manufacturer Part Number: 122910

UNSPSC: 47131600

Code: NEW

Additional Details:
Additional Details: Weight: 2.10000kg



Product Code. 15273546

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

For Use With (Application) For use on floors, walls, and ceilings in cleanroom environments. Ideal for the pull-and-lift cleaning technique
Material Aluminum
Product Type Aluminum Extendable Mop with Sponge Mop Head, Straight
For Use With (Equipment) Roll-O-matic self wringing cleanroom mops
Includes Color coding tabs and ergonomic grip, extendable handle
Autoclavable No
Color Blue