Learn More
Labconco™ FreeZone Benchtop Freeze Dry System
Brand:  Labconco™ 7670560F
Code : NEW
Additional Details : Weight : 101.00000kg
 View more versions of this product
Product Code. 15370461
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
The 2. 5 liter FreeZone™ Benchtop freeze dry system offers a simple to use, space saving design in an economical and versatile configuration. Labconco's exclusive “wave graphs” communicate both temperature and vacuum information. The integral vacuum control valve provides precise control of the vacuum level to optimise the drying process. An exclusive moisture sensor in the condenser drain tubing prevents accidental pump oil contamination from liquids left over from previous use. This configuration includes a clear drying chamber with eight valve ports so that drying can be performed on the included shelves or in flasks. A rotary vane vacuum pump capable of 0.002mbar is required.
  • Simple and logical controls that are easy to use with an LCD display screen
  • Integrated vacuum control
  • Compact design that is designed to integrate into the laboratory workflow
  • Audible and visual alarms to warn if an upset in the drying conditions exists
  • RS232 data output
Videos