Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, 5 in/127 mm
Product Code. 15773225
Product Code. 15773225
Brand: Eppendorf™ 6001000510
Only null left
Add to basket
Add to basket
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, 5 in/127 mm, side access, stainless steel- Rack for 127 mm/5.0 in boxes (15/50 mL conical tubes)- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 141 x 141 x 681 (5.6 x 5.6 x 26.8 in)- Max. box number per rack: 5 (5 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 32 racks), Innova C760 (max. 40 racks), CryoCube™ FC660 series (max. 40 racks)- For ULT freezer: Innova C580 (max. 160 boxes), Innova C760 (max. 200 boxes), CryoCube™ FC660 series (max. 200 boxes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack