Learn More
Fraction V Bovine Albumin, pH 7, Thermo Scientific Chemicals
Heat-Shock fractionated
Brand: Thermo Scientific J10857.A1
Additional Details : CAS Number : 9048-46-8 Weight : 2.00000kg
Description
- Source: Bovine blood
- Form: Lyophilized powder
- Assay (electrophoresis): ≥98.0%
- Protein: ≥96.0%
- pH (2%, H2O): 6.5 - 7.5
- Loss On Drying: ≤5.0%
- Residue on Ignition: ≤2.0%
- CAS: 9048-46-8
Specifications
6.5 to 7.5 | |
Bovine blood | |
Plastic bottle | |
Lyophilized |
1 kg | |
4°C | |
Fraction V Bovine Albumin |
RUO – Research Use Only