missing translation for 'onlineSavingsMsg'
Learn More

FR4, PE-Cyanine7, clone: eBio12A5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-5445-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15320970

  • £105.68 / 25µg


EU Stock 2
Add to basket

Description

Description

Description: The monoclonal antibody eBio12A5 recognizes FR4, also known as Folate receptor 4, FR delta and FBP3 (folate binding protein3). FR4 is a heavily glycosylated, 35 kD receptor for folic acid and the physiologic circulating form of the vitamin, N5-methyltetrahydrofolate. Natural Treg have high levels of FR4 and together with CD25 can be used to distinguish natural Treg, effector T cells, memory-like T cells and naive T cells. FR4 high CD25 high cells are natural Treg with high levels of Foxp3. FR4 high CD25 low cells are identified as central memory T cells, which upon stimulation, can proliferate and produce large amounts of IL-2. In contrast, FR4 low CD25 high cells secrete proinflammatory cytokines such as IFN gamma and IL-17 and have been characterized as effector memory T cells.Based on co-staining studies, the epitopes recognized by eBioTH6 and eBio12A5 are different, thereby allowing functional studies with eBioTH6 to be evaluated with eBio12A5.Applications Reported: This eBio12A5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio12A5 antibody has been tested by flow cytyometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

FR4
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9EQF4
Folate receptor 4, FOLR4, FRdelta, FRd
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBio12A5
PE-Cyanine7
Izumo1r
Conjugated
Izumo1r
IgG2b, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
64931
Videos

For Research Use Only.